missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylbutyryl chloride, 99%, Thermo Scientific Chemicals

Catalog Number 10742073
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2736-40-5
C6H11ClO
134.60
MFCD00018812
SMUKODJVMQOSAB-UHFFFAOYSA-N
2-ethylbutyryl chloride, 2-ethylbutyroyl chloride, butanoyl chloride, 2-ethyl, butyryl chloride, 2-ethyl, 2-ethylbutyrylchloride, diethylacetylchloride, diethylacetyl chloride, acmc-1cqal, 2-ethyl butyryl chloride, 2-ethyl-butyryl chloride
75954
2-ethylbutanoyl chloride
CCC(CC)C(Cl)=O
This item is not returnable. View return policy
2736-40-5
C6H11ClO
134.60
MFCD00018812
SMUKODJVMQOSAB-UHFFFAOYSA-N
2-ethylbutyryl chloride, 2-ethylbutyroyl chloride, butanoyl chloride, 2-ethyl, butyryl chloride, 2-ethyl, 2-ethylbutyrylchloride, diethylacetylchloride, diethylacetyl chloride, acmc-1cqal, 2-ethyl butyryl chloride, 2-ethyl-butyryl chloride
75954
2-ethylbutanoyl chloride
CCC(CC)C(Cl)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 0.9820g/mL
Boiling Point 138°C
Flash Point 33°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (C2H5)2CHCOCl
Refractive Index 1.4230 to 1.425
Quantity 5 g
Specific Gravity 0.982
Formula Weight 134.61
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Ethylbutyryl chloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Ethylbutyryl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'