missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylaniline, 98%, Thermo Scientific Chemicals

Catalog Number p-126879
Click to view available options
Quantity:
5 g
250 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
578-54-1
C8H11N
121.18
MFCD00007751
MLPVBIWIRCKMJV-UHFFFAOYSA-N
o-ethylaniline, benzenamine, 2-ethyl, aniline, o-ethyl, 2-ethylbenzenamine, o-aminoethylbenzene, aniline, 2-ethyl, 2-ethyl aniline, 2-ethylphenylamine, unii-gr557n47k6, ccris 2858
11357
2-ethylaniline
CCC1=CC=CC=C1N
This item is not returnable. View return policy
578-54-1
C8H11N
121.18
MFCD00007751
MLPVBIWIRCKMJV-UHFFFAOYSA-N
o-ethylaniline, benzenamine, 2-ethyl, aniline, o-ethyl, 2-ethylbenzenamine, o-aminoethylbenzene, aniline, 2-ethyl, 2-ethyl aniline, 2-ethylphenylamine, unii-gr557n47k6, ccris 2858
11357
2-ethylaniline
CCC1=CC=CC=C1N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point -44°C
Color Brown-Red to Yellow
Density 0.9800g/mL
Boiling Point 210°C
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula C2H5C6H4NH2
Refractive Index 1.5590 to 1.561
Quantity 5 g
Beilstein 12, II, 584
Specific Gravity 0.98
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and toluene
Formula Weight 121.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Ethylaniline, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Ethylaniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'