missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Ethylacrolein, tech. 90%, stab. with 50ppm hydroquinone, Thermo Scientific Chemicals
28.60 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
100 g
Description
2-Ethylacrolein has been used in total synthesis of (±)-tabersonine, Aspidosperma alkaloid and synthesis of conjugate with bovine serum albumin tethered to Tn.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Ethylacrolein has been used in total synthesis of (+/-)-tabersonine, Aspidosperma alkaloid and synthesis of conjugate with bovine serum albumin tethered to Tn.
Solubility
Soluble in alcohol. Insoluble in water.
Notes
Air sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store at 2-8°C.
2-Ethylacrolein has been used in total synthesis of (+/-)-tabersonine, Aspidosperma alkaloid and synthesis of conjugate with bovine serum albumin tethered to Tn.
Solubility
Soluble in alcohol. Insoluble in water.
Notes
Air sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store at 2-8°C.
Specifications
Density | 0.859 |
Boiling Point | 92°C to 93°C |
Flash Point | 1°C (34°F) |
Odor | Pungent |
Refractive Index | 1.4245 |
Quantity | 5 g |
UN Number | UN1992 |
Beilstein | 1098378 |
Sensitivity | Air sensitive |
Solubility Information | Soluble in alcohol. Insoluble in water. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'