missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-2-oxazoline, 99+%, Thermo Scientific Chemicals

Catalog Number p-4510375
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
10431-98-8
C5H9NO
99.13
MFCD00005302
NYEZZYQZRQDLEH-UHFFFAOYSA-N
2-ethyl-2-oxazoline, 2-ethyloxazoline, 2-ethyl-4,5-dihydrooxazole, oxazole, 2-ethyl-4,5-dihydro, 2-oxazoline, 2-ethyl, unii-b8cd92t4p5, unii-jm7uh9q3yn, unii-hnx7574gtx, unii-s5dpg6k67i, ethyl oxazoline
66412
2-ethyl-4,5-dihydro-1,3-oxazole
CCC1=NCCO1
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
10431-98-8
C5H9NO
99.13
MFCD00005302
NYEZZYQZRQDLEH-UHFFFAOYSA-N
2-ethyl-2-oxazoline, 2-ethyloxazoline, 2-ethyl-4,5-dihydrooxazole, oxazole, 2-ethyl-4,5-dihydro, 2-oxazoline, 2-ethyl, unii-b8cd92t4p5, unii-jm7uh9q3yn, unii-hnx7574gtx, unii-s5dpg6k67i, ethyl oxazoline
66412
2-ethyl-4,5-dihydro-1,3-oxazole
CCC1=NCCO1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -62.0°C
Density 0.9800g/mL
Boiling Point 128.4°C
Flash Point 29°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.4360 to 1.4380
Quantity 100 g
Specific Gravity 0.98
Solubility Information Solubility in water: miscible
Viscosity 0.88 mPa.s (20°C)
Formula Weight 99.13
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 2-Ethyl-2-oxazoline
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Ethyl-2-oxazoline
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'