missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-2-(hydroxymethyl)-1,3-propanediol, 98%, Thermo Scientific Chemicals

Catalog Number p-3736450
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
This item is not returnable. View return policy
77-99-6
C6H14O3
134.18
MFCD00004694
ZJCCRDAZUWHFQH-UHFFFAOYSA-N
trimethylolpropane, 2-ethyl-2-hydroxymethyl propane-1,3-diol, ethriol, 2-ethyl-2-hydroxymethyl-1,3-propanediol, hexaglycerine, ettriol, etriol, trimethylol propane, tmp alcohol, 1,1,1-tris hydroxymethyl propane
6510
2-ethyl-2-(hydroxymethyl)propane-1,3-diol
CCC(CO)(CO)CO
This item is not returnable. View return policy
77-99-6
C6H14O3
134.18
MFCD00004694
ZJCCRDAZUWHFQH-UHFFFAOYSA-N
trimethylolpropane, 2-ethyl-2-hydroxymethyl propane-1,3-diol, ethriol, 2-ethyl-2-hydroxymethyl-1,3-propanediol, hexaglycerine, ettriol, etriol, trimethylol propane, tmp alcohol, 1,1,1-tris hydroxymethyl propane
6510
2-ethyl-2-(hydroxymethyl)propane-1,3-diol
CCC(CO)(CO)CO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Acidity 0.002% max. (as formic acid)
CAS Min % 97.5
CAS Max % 100.0
Melting Point 56.0°C to 60.0°C
Color White
Boiling Point 160°C (0.7 hPa)
Flash Point 172°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Linear Formula C2H5C(CH2OH)3
Quantity 500 g
Beilstein 01, III, 2349
Solubility Information Solubility in water: soluble. Other solubilities: freely soluble in alcohols,insoluble in hydrocarbons
Viscosity 157 mPa.s (75°C)
Formula Weight 134.18
Percent Purity 98%
Physical Form Flakes
Chemical Name or Material 2-Ethyl-2-(hydroxymethyl)-1, 3-propanediol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'