missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxyphenol, 98%, Thermo Scientific Chemicals

Catalog Number 10571882
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
94-71-3
C8H10O2
138.17
MFCD00002187
MOEFFSWKSMRFRQ-UHFFFAOYSA-N
o-ethoxyphenol, phenol, 2-ethoxy, guaethol, guethol, 2-ethyloxyphenol, guaiethol, pyrocatechol monoethyl ether, catechol monoethyl ether, phenol, o-ethoxy, unii-878iw8p9pw
66755
2-ethoxyphenol
CCOC1=CC=CC=C1O
This item is not returnable. View return policy
94-71-3
C8H10O2
138.17
MFCD00002187
MOEFFSWKSMRFRQ-UHFFFAOYSA-N
o-ethoxyphenol, phenol, 2-ethoxy, guaethol, guethol, 2-ethyloxyphenol, guaiethol, pyrocatechol monoethyl ether, catechol monoethyl ether, phenol, o-ethoxy, unii-878iw8p9pw
66755
2-ethoxyphenol
CCOC1=CC=CC=C1O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 20.0°C to 25.0°C
Color Colorless to Yellow
Density 1.0900g/mL
Boiling Point 216.0°C to 217.0°C
Flash Point 91°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C2H5OC6H4OH
Refractive Index 1.5280 to 1.5300
Quantity 100 g
Specific Gravity 1.09
Formula Weight 138.17
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Ethoxyphenol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Ethoxyphenol
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H412-Harmful to aquatic life with long lasting effects.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'