missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxynaphthoic acid, 98%, Thermo Scientific™

Catalog Number p-126000
Click to view available options
Quantity:
10 g
This item is not returnable. View return policy
2224-00-2
C13H11O3
215.23
MFCD00004008
MYFBSSDLYGWAHH-UHFFFAOYSA-M
2-ethoxy-1-naphthoic acid, 2-ethoxynaphthoic acid, 1-naphthalenecarboxylic acid, 2-ethoxy, 2-ethoxy-1-naphthoicacid, 2-ethoxynaphthalenecarboxylic acid, acmc-209ftv, rarechem al be 0566, timtec-bb sbb005771, 2-ethoxynaphtoic acid
75210
CCOC1=CC=C2C=CC=CC2=C1C([O-])=O
This item is not returnable. View return policy
2224-00-2
C13H11O3
215.23
MFCD00004008
MYFBSSDLYGWAHH-UHFFFAOYSA-M
2-ethoxy-1-naphthoic acid, 2-ethoxynaphthoic acid, 1-naphthalenecarboxylic acid, 2-ethoxy, 2-ethoxy-1-naphthoicacid, 2-ethoxynaphthalenecarboxylic acid, acmc-209ftv, rarechem al be 0566, timtec-bb sbb005771, 2-ethoxynaphtoic acid
75210
CCOC1=CC=C2C=CC=CC2=C1C([O-])=O

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 141°C to 145°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 10 g
IUPAC Name 2-ethoxynaphthalene-1-carboxylic acid
Formula Weight 216.24
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Ethoxynaphthoic acid, 98%
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'