missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxyethyl Ether, 98+%, Extra Pure, Thermo Scientific Chemicals

Catalog Number p-3736417
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
10 kg
Packaging:
Glass bottle
Metal drum
This item is not returnable. View return policy
112-36-7
MFCD00009254
RRQYJINTUHWNHW-UHFFFAOYSA-N
diethylene glycol diethyl ether, 2-ethoxyethyl ether, diethyl carbitol, ethyl diglyme, 1-ethoxy-2-2-ethoxyethoxy ethane, bis 2-ethoxyethyl ether, diethyldiethylene glycol, 3,6,9-trioxaundecane, degdee, ether, bis 2-ethoxyethyl
8179
CHEBI:44664
1-ethoxy-2-(2-ethoxyethoxy)ethane
CCOCCOCCOCC
This item is not returnable. View return policy
112-36-7
MFCD00009254
RRQYJINTUHWNHW-UHFFFAOYSA-N
diethylene glycol diethyl ether, 2-ethoxyethyl ether, diethyl carbitol, ethyl diglyme, 1-ethoxy-2-2-ethoxyethoxy ethane, bis 2-ethoxyethyl ether, diethyldiethylene glycol, 3,6,9-trioxaundecane, degdee, ether, bis 2-ethoxyethyl
8179
CHEBI:44664
1-ethoxy-2-(2-ethoxyethoxy)ethane
CCOCCOCCOCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.0
CAS Max % 100.0
Density 0.9g/mL
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Molecular Formula C8H18O3
Linear Formula (C2H5OCH2CH2)2O
Refractive Index 1.4110 to 1.4130 (20°C, 589nm)
Quantity 500 g
Beilstein 01, II, 519
Residue after Evaporation 0.05% max.
Merck Index 15, 3132
Molecular Weight (g/mol) 162.229
Formula Weight 162.23
Percent Purity ≥98.0%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material 2-Ethoxyethyl ether
Show More Show Less
Product Identifier
  • 2-Ethoxyethyl ether

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'