missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxyethanol, 99%, extra pure, Thermo Scientific Chemicals

Catalog Number p-3736423
Click to view available options
Quantity:
1 L
2.5 L
25 L
Packaging:
Glass Bottle
Metal drum
This item is not returnable. View return policy
110-80-5
C4H10O2
90.12
MFCD00002869
ZNQVEEAIQZEUHB-UHFFFAOYSA-N
cellosolve, oxitol, ethyl cellosolve, ethylene glycol monoethyl ether, ethanol, 2-ethoxy, ethyl glycol, emkanol, hydroxy ether, ethylene glycol ethyl ether, dowanol ee
8076
CHEBI:46788
2-ethoxyethanol
CCOCCO
This item is not returnable. View return policy
110-80-5
C4H10O2
90.12
MFCD00002869
ZNQVEEAIQZEUHB-UHFFFAOYSA-N
cellosolve, oxitol, ethyl cellosolve, ethylene glycol monoethyl ether, ethanol, 2-ethoxy, ethyl glycol, emkanol, hydroxy ether, ethylene glycol ethyl ether, dowanol ee
8076
CHEBI:46788
2-ethoxyethanol
CCOCCO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point -70°C
Density 0.9300g/mL
Boiling Point 135.0°C
Flash Point 40°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Linear Formula C2H5OCH2CH2OH
Refractive Index 1.4060 to 1.4100
Quantity 1 L
Beilstein 01, 467
Merck Index 15, 3803
Specific Gravity 0.93
Solubility Information Solubility in water: complete solubility. Other solubilities: miscible with most organic solvents
Viscosity 2.08 mPa.s (20°C)
Formula Weight 90.12
Percent Purity 99%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material 2-Ethoxyethanol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Ethoxyethanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Flammable liquid Category 3
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.
  • H331-Toxic if inhaled.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'