missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxyethanol, 99%, Thermo Scientific Chemicals

Catalog Number 11463127
Click to view available options
Quantity:
500 mL
2500 mL
This item is not returnable. View return policy
110-80-5
C4H10O2
90.12
MFCD00002869
ZNQVEEAIQZEUHB-UHFFFAOYSA-N
cellosolve, oxitol, ethyl cellosolve, ethylene glycol monoethyl ether, ethanol, 2-ethoxy, ethyl glycol, emkanol, hydroxy ether, ethylene glycol ethyl ether, dowanol ee
8076
CHEBI:46788
2-ethoxyethan-1-ol
CCOCCO
This item is not returnable. View return policy
110-80-5
C4H10O2
90.12
MFCD00002869
ZNQVEEAIQZEUHB-UHFFFAOYSA-N
cellosolve, oxitol, ethyl cellosolve, ethylene glycol monoethyl ether, ethanol, 2-ethoxy, ethyl glycol, emkanol, hydroxy ether, ethylene glycol ethyl ether, dowanol ee
8076
CHEBI:46788
2-ethoxyethan-1-ol
CCOCCO

A widely used solvent for nitrocelluloses and dyes2-Ethoxyethanol is widely used as a varnish remover and degreasing solution. It is a solvent used for nitrocellulose. It is used as a component for dyes, inks, cleaning agents, resins, paints, destaining solution for lipoproteins on polyacrylamide gels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A widely used solvent for nitrocelluloses and dyes2-Ethoxyethanol is widely used as a varnish remover and degreasing solution. It is a solvent used for nitrocellulose. It is used as a component for dyes, inks, cleaning agents, resins, paints, destaining solution for lipoproteins on polyacrylamide gels.

Solubility
Miscible with water, acetone, ethyl acetate, benzene, carbon tetrachloride, diethyl ether, methanol, hydrocarbons and ethanol.

Notes
Incompatible with strong acids, strong bases, strong oxidizing agents, aluminum, alkalies and copper.
TRUSTED_SUSTAINABILITY
Melting Point -90°C
Density 0.931
Boiling Point 134°C to 135°C
Flash Point 44°C (111°F)
Odor Ether-like
Linear Formula CH3CH2OCH2CH2OH
Refractive Index 1.4085
Quantity 500 mL
UN Number UN1171
Beilstein 1098271
Merck Index 14,3750
Solubility Information Miscible with water,acetone,ethyl acetate,benzene,carbon tetrachloride,diethyl ether,methanol,hydrocarbons and ethanol.
Formula Weight 90.12
Percent Purity 99%
Chemical Name or Material 2-Ethoxyethanol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Ethoxyethanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 3
  • Reproductive toxicity Category 1B
  • Acute toxicity Category 4
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H331-Toxic if inhaled.
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P201-Obtain special instructions before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'