missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Ethoxycarbonyl-4-fluorobenzeneboronic acid, 97%

Brand:  Alfa Aesar™ H61322.03

Code : NEW

Additional Details : CAS Number : 1072951-40-6 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15457848

  • £74.62 / 1g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

2-Ethoxycarbonyl-4-fluorobenzeneboronic acid
211.98
2-Ethoxycarbonyl-4-fluorophenylboronic acid
1072951-40-6
MFCD11053864

Specifications

2-Ethoxycarbonyl-4-fluorobenzeneboronic acid
1072951-40-6
97%
MFCD11053864
1g
Solid
211.98
2-Ethoxycarbonyl-4-fluorophenylboronic acid
Videos
SDS