missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Ethoxybenzoic acid, 98%, Thermo Scientific Chemicals
13.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
500 g
Description
2-Ethoxybenzoic acid is used in thermally induced solvent-free reactions of triphenylbismuth with a series of thiols and carboxylic acids. It serves as an active component of dental cement. Further, it prevents the entry of root-canal pathogens into periapical regions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Ethoxybenzoic acid is used in thermally induced solvent-free reactions of triphenylbismuth with a series of thiols and carboxylic acids. It serves as an active component of dental cement. Further, it prevents the entry of root-canal pathogens into periapical regions.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
2-Ethoxybenzoic acid is used in thermally induced solvent-free reactions of triphenylbismuth with a series of thiols and carboxylic acids. It serves as an active component of dental cement. Further, it prevents the entry of root-canal pathogens into periapical regions.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Melting Point | 19°C to 20°C |
Density | 1.105 |
Boiling Point | 174°C to 175°C (15 mmHg) |
Flash Point | >110°C (230°F) |
Refractive Index | 1.541 |
Quantity | 25 g |
Beilstein | 2691039 |
Solubility Information | Soluble in water. |
Formula Weight | 166.18 |
Percent Purity | 98% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'