missing translation for 'onlineSavingsMsg'
Learn More

2-(Diphenylphosphino)benzoic Acid, 98%, ACROS Organics™

Brand:  Acros Organics 382930010

Code : 29

Additional Details : CAS Number : 17261-28-8 Weight : 0.05100kg


Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 10566363

  • POA / 1g

This item cannot be ordered online because the price is subject to change. Please contact our Product Support team on 01509 555500 for assistance.



Specifications

Chemical Identifiers

2-(Diphenylphosphino)benzoic acid
97.5
98%
C19H15O2P
306.3
17261-28-8
100.0
98%
(C6H5)2PC6H4COOH
MFCD00674024

Specifications

2-(Diphenylphosphino)benzoic acid
177.0° to 181.0°C
17261-28-8
100.0
Glass bottle
98%
C19H15O2P
306.3
Light yellow
1g
97.5
Authentic
Powder
98%
(C6H5)2PC6H4COOH
MFCD00674024
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-(Diphenylphosphino)benzoic acid

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS