missing translation for 'onlineSavingsMsg'
Learn More

2-(Dimethylaminomethyl)-1-cyclohexanone hydrochloride, 98%, Thermo Scientific Chemicals

Catalog Number 10449412
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
42036-65-7
C9H17NO·HCl
191.7
MFCD00102580
CLVHTSWMNNSUSH-UHFFFAOYSA-N
2-dimethylamino methyl cyclohexanone hydrochloride, 2-dimethylaminomethyl-1-cyclohexanone hydrochloride, dimethyl 2-oxocyclohexyl methyl ammonium chloride, 2-dimethylamino methyl cyclohexan-1-one hydrochloride, 2-dimethylaminomethyl-cyclohexanone, 2-dimethylamino methyl cyclohexan-1-one, chloride, tramadol related compound b, 2-dimethylaminomethyl-cyclohexanone hydrochloride, tramadol related compound b usp
359482
2-[(dimethylamino)methyl]cyclohexan-1-one;hydrochloride
CN(C)CC1CCCCC1=O.Cl
This item is not returnable. View return policy
42036-65-7
C9H17NO·HCl
191.7
MFCD00102580
CLVHTSWMNNSUSH-UHFFFAOYSA-N
2-dimethylamino methyl cyclohexanone hydrochloride, 2-dimethylaminomethyl-1-cyclohexanone hydrochloride, dimethyl 2-oxocyclohexyl methyl ammonium chloride, 2-dimethylamino methyl cyclohexan-1-one hydrochloride, 2-dimethylaminomethyl-cyclohexanone, 2-dimethylamino methyl cyclohexan-1-one, chloride, tramadol related compound b, 2-dimethylaminomethyl-cyclohexanone hydrochloride, tramadol related compound b usp
359482
2-[(dimethylamino)methyl]cyclohexan-1-one;hydrochloride
CN(C)CC1CCCCC1=O.Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 147°C to 151°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in methanol,ethanol
Formula Weight 191.7
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-(Dimethylaminomethyl)-1-cyclohexanone hydrochloride, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'