missing translation for 'onlineSavingsMsg'
Learn More

2-Dimethylaminoisopropyl chloride hydrochloride, 98.5%, ACROS Organics™

Brand:  Acros Organics 115831000

Code : 29

Additional Details : CAS Number : 4584-49-0 Weight : 0.15000kg

Quantity 100g
Packaging Glass bottle

 View more versions of this product

Product Code. 10466720

  • £11.70 / 100g


EU Stock 9
Add to basket

Specifications

Chemical Identifiers

2-Dimethylaminoisopropyl chloride hydrochloride
4584-49-0
100.0
98.5%
CH3CH(Cl)CH2N(CH3)2·HCl
MFCD00012534
98.5%
98.0
98.5%
C5H12ClN·HCl
158.07
2-Chloro-N, N-dimethylpropylamine hydrochloride, DMIC

Specifications

2-Dimethylaminoisopropyl chloride hydrochloride
191.0° to 195.0°C
98.5%
98.0
1% max.
Glass bottle
Crystalline Powder
98.5%
CH3CH(Cl)CH2N(CH3)2·HCl
MFCD00012534
White to light cream
100g
4584-49-0
100.0
Authentic
Solubility in water: 2000g/L (20°C). Other solubilities: soluble in methanol, insoluble in benzene and toluene
98.5%
C5H12ClN·HCl
158.07
2-Chloro-N, N-dimethylpropylamine hydrochloride, DMIC
Videos
SDS