missing translation for 'onlineSavingsMsg'
Learn More

(2-Dimethylaminoethyl)triphenylphosphonium bromide, 97%, Thermo Scientific™

Catalog Number p-115248
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
21331-80-6
C22H25BrNP
414.33
MFCD00011794
YIIHEMGEQQWSMA-UHFFFAOYSA-M
2-dimethylaminoethyl triphenylphosphonium bromide, 2-dimethylamino ethyl triphenylphosphonium bromide, 2-dimethylaminoethyltriphenylphosphonium bromide, 2-dimethylaminoethyl triphenylphosphoniumbromide, 2-dimethylamino ethyl triphenylphosphanium bromide, acmc-1ci7c, triphenyldimethylaminoethylphosphonium bromide, 2-dimethylaminoethyltriphenylphosphoniumbromide
2724136
[Br-].CN(C)CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
21331-80-6
C22H25BrNP
414.33
MFCD00011794
YIIHEMGEQQWSMA-UHFFFAOYSA-M
2-dimethylaminoethyl triphenylphosphonium bromide, 2-dimethylamino ethyl triphenylphosphonium bromide, 2-dimethylaminoethyltriphenylphosphonium bromide, 2-dimethylaminoethyl triphenylphosphoniumbromide, 2-dimethylamino ethyl triphenylphosphanium bromide, acmc-1ci7c, triphenyldimethylaminoethylphosphonium bromide, 2-dimethylaminoethyltriphenylphosphoniumbromide
2724136
[Br-].CN(C)CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 201.0°C
Color Brown-Yellow to White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula (CH3)2NCH2CH2P(C6H5)3Br
Quantity 10 g
Fieser 08,184; 09,291; 13,174; 15,198; 16,205
IUPAC Name 2-(dimethylamino)ethyl-triphenylphosphanium;bromide
Formula Weight 414.32
Percent Purity 97%
Physical Form Powder
Chemical Name or Material (2-Dimethylaminoethyl)triphenylphosphonium bromide, 97%
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'