missing translation for 'onlineSavingsMsg'
Learn More

2-Dimethylaminoethyl Chloride Hydrochloride 99%, ACROS Organics™

Brand:  Acros Organics 115820025

Code : 29

Additional Details : CAS Number : 4584-46-7 Weight : 2.50000kg Transport : UN number : 3077 Chem class : 9 Pack group : III


Quantity 2.5kg
Packaging Plastic bottle

 View more versions of this product

Product Code. 10157693

  • £136.50 / 2.50kg


EU Stock 1
Add to basket


Description

Description

Alkylation reagent
Specifications

Chemical Identifiers

2-Dimethylaminoethyl chloride hydrochloride
107-99-3
100.0
99%
(CH3)2NCH2CH2Cl·HCl
MFCD00012516
99%
98.5
99%
C4H10ClN·HCl
144.05
2-Chloro-N, N-dimethylethylamine hydrochloride

Specifications

2-Dimethylaminoethyl chloride hydrochloride
203.0° to 210.0°C
99%
48.4 to 50.0% (total chloride), 24.1 to 25.1% (ionic chloride)
100.0
Plastic bottle
Crystalline Powder or Crystals
99%
1% Max. (K.F.)
144.05
04, 133
White to brown-yellow
2.5kg
107-99-3
98.5
Authentic
Solubility in water: 2000g/L (20°C). Other solubilities: soluble in alcohol (20°C)
99%
C4H10ClN·HCl
(CH3)2NCH2CH2Cl·HCl
MFCD00012516
2-Chloro-N, N-dimethylethylamine hydrochloride
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Dimethylaminoethyl chloride hydrochloride

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Germ cell mutagenicity Category 2
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H341-Suspected of causing genetic defects.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS