missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Dimethylaminoethyl chloride hydrochloride, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent
Specifications
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Melting Point | 203.0°C to 210.0°C |
Color | Brown-Yellow to White |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% |
Packaging | Plastic bottle |
Linear Formula | (CH3)2NCH2CH2Cl·HCl |
Quantity | 500 g |
Beilstein | 04, 133 |
Show More |
Safety and Handling
Product Identifier
- 2-Dimethylaminoethyl chloride hydrochloride
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Germ cell mutagenicity Category 2
- Specific target organ toxicity after repeated exposure Category 2
- H341-Suspected of causing genetic defects.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
- H302-Harmful if swallowed.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'