missing translation for 'onlineSavingsMsg'
Learn More

2-Dimethylaminoethyl acetate, 98%, Thermo Scientific Chemicals

Catalog Number p-115467
Click to view available options
Quantity:
25 g
This item is not returnable. View return policy
1421-89-2
C6H13NO2
131.18
MFCD00041912
GOLSFPMYASLXJC-UHFFFAOYSA-N
2-dimethylamino ethyl acetate, dimethylaminoethyl acetate, dimethylaminoethanol acetate, 2-dimethylaminoethanol acetate, acetic acid, 2-dimethylamino ethyl ester, acetic acid 2-dimethylaminoethyl ester, 2-dimethylamino ethanol acetic ester, acmc-1c2gl, 2-dimethylaminoethyl=acetate, 2-dimethylamino ethylacetate
15013
2-(dimethylamino)ethyl acetate
CN(C)CCOC(C)=O
This item is not returnable. View return policy
1421-89-2
C6H13NO2
131.18
MFCD00041912
GOLSFPMYASLXJC-UHFFFAOYSA-N
2-dimethylamino ethyl acetate, dimethylaminoethyl acetate, dimethylaminoethanol acetate, 2-dimethylaminoethanol acetate, acetic acid, 2-dimethylamino ethyl ester, acetic acid 2-dimethylaminoethyl ester, 2-dimethylamino ethanol acetic ester, acmc-1c2gl, 2-dimethylaminoethyl=acetate, 2-dimethylamino ethylacetate
15013
2-(dimethylamino)ethyl acetate
CN(C)CCOC(C)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Density 0.9200g/mL
Boiling Point 152°C to 154°C
Flash Point 47°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3COOCH2CH2N(CH3)2
Refractive Index 1.4155 to 1.4175
Quantity 25 g
Specific Gravity 0.92
Formula Weight 131.17
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Dimethylaminoethyl acetate, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Dimethylaminoethyl acetate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'