missing translation for 'onlineSavingsMsg'
Learn More

2-Diethylaminoethylchloride hydrochloride, 99.5%, ACROS Organics™

Brand:  Acros Organics 114030025

Code : 29

Additional Details : CAS Number : 869-24-9 Weight : 2.55000kg Transport : UN number : 2928 Chem class : 1a Pack group : II


Quantity 2.5kg
Packaging Plastic drum

 View more versions of this product

Product Code. 11384906

  • £147.30 / 2.50kg


EU Stock 1
Add to basket


Specifications

Chemical Identifiers

2-Diethylaminoethylchloride hydrochloride
99.4
99.5%
C6H14ClN·HCl
172.1
2-Chloro-N, N-diethylethylamine hydrochloride
869-24-9
100.0
99.5%
(C2H5)2NCH2CH2Cl·HCl
MFCD00012519

Specifications

2-Diethylaminoethylchloride hydrochloride
99.4
100.0
Plastic drum
Crystalline Powder
208.0° to 212.0°C
2.5kg
(C2H5)2NCH2CH2Cl·HCl
MFCD00012519
2-Chloro-N, N-diethylethylamine hydrochloride
869-24-9
Authentic
1% max. (105°C, 3 hrs)
White to slightly beige
99.5%
99.5%
C6H14ClN·HCl
172.1
04, III, 240
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Diethylaminoethylchloride hydrochloride

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H300-Fatal if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/A§.
  • P330-Rinse mouth.

SDS