2-Diethylaminoethylchloride hydrochloride, 99.5%, ACROS Organics™


Overview
Brand: Acros Organics

Manufacturer Part Number: 114030010

UNSPSC: 12350000

Code: 29

Additional Details:
Additional Details: CAS Number: 869-24-9 Weight: 1.05000kg Transport: UN number : 2928 Chem class : 1a Pack group : II


Packaging Glass Bottle
Quantity 1kg

Product Code. 10122380

Quantity Price
1 £ 72.5 / 1kg
Estimated Shipment
22-03-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 2-Diethylaminoethylchloride hydrochloride
CAS 869-24-9
CAS Min % 99.4
CAS Max % 100.0
Assay ≥99.4% (on dry substance)
Assay Percent Range 99.5%
Molecular Formula C6H14ClN·HCl
Linear Formula (C2H5)2NCH2CH2Cl·HCl
Formula Weight 172.1
MDL Number MFCD00012519
Synonym 2-Chloro-N, N-diethylethylamine hydrochloride

Specification

Assay ≥99.4% (on dry substance)
Formula Weight 172.1
Physical Form Crystalline Powder
Quantity 1kg
Color White to slightly beige
Beilstein 04, III, 240
Packaging Glass Bottle
CAS Min % 99.4
Linear Formula (C2H5)2NCH2CH2Cl·HCl
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
CAS 869-24-9
Loss on Drying 1% max. (105°C, 3 hrs)
Assay Percent Range 99.5%
MDL Number MFCD00012519
Health Hazard 2 GHS H Statement:
Fatal if swallowed.
Causes serious eye damage.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Toxic to aquatic life with long lasting effects.
Molecular Formula C6H14ClN·HCl
CAS Max % 100.0
Health Hazard 1 GHS Signal Word: Danger
Chemical Name or Material 2-Diethylaminoethylchloride hydrochloride
Synonym 2-Chloro-N, N-diethylethylamine hydrochloride
Melting Point 208° to 212°C
Documentation

Technical documents

Safety and Handling

GHS Signal Word: Danger

GHS H Statement:
Fatal if swallowed.
Causes serious eye damage.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Toxic to aquatic life with long lasting effects.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.