missing translation for 'onlineSavingsMsg'
Learn More

2-Diethylaminoethylchloride hydrochloride, 99.5%, ACROS Organics™

Brand:  Acros Organics 114030050

Code : 29

Additional Details : CAS Number : 869-24-9 Weight : 0.05500kg Transport : UN number : 2928 Chem class : 6.1 Pack group : II


Packaging Glass Bottle
Quantity 5g

 View more versions of this product

Product Code. 10050830

  • £11.32 / 5g

Estimated Shipment: 19-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

869-24-9
172.09
RAGSWDIQBBZLLL-UHFFFAOYSA-N
13363
[H+].[Cl-].CCN(CC)CCCl
C6H15Cl2N
MFCD00012519
2-chloro-n,n-diethylethanamine hydrochloride, 2-diethylaminoethylchloride hydrochloride, 2-chlorotriethylamine hydrochloride, 2-chloro-n,n-diethylethylamine hydrochloride, 2-diethylamino ethyl chloride hydrochloride, diethylaminoethyl chloride hydrochloride, n,n-diethyl-2-chloroethylamine hydrochloride, 2-chloroethyl diethylamine hydrochloride, unii-ycy6eqt3wa, ethanamine, 2-chloro-n,n-diethyl-, hydrochloride
2-chloro-N,N-diethylethanamine;hydrochloride

Specifications

2-Diethylaminoethylchloride hydrochloride
1% max. (105°C, 3 hrs)
White to Beige
5g
99.4
99.5%
(C2H5)2NCH2CH2Cl·HCl
04, III, 240
Solubility in water: 2000g/L (20°C)
[H+].[Cl-].CCN(CC)CCCl
172.09
172.1
99.5%
Authentic
Glass Bottle
208.0°C to 212.0°C
869-24-9
100.0
C6H15Cl2N
MFCD00012519
2-chloro-n,n-diethylethanamine hydrochloride, 2-diethylaminoethylchloride hydrochloride, 2-chlorotriethylamine hydrochloride, 2-chloro-n,n-diethylethylamine hydrochloride, 2-diethylamino ethyl chloride hydrochloride, diethylaminoethyl chloride hydrochloride, n,n-diethyl-2-chloroethylamine hydrochloride, 2-chloroethyl diethylamine hydrochloride, unii-ycy6eqt3wa, ethanamine, 2-chloro-n,n-diethyl-, hydrochloride
RAGSWDIQBBZLLL-UHFFFAOYSA-N
2-chloro-N,N-diethylethanamine;hydrochloride
13363
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Diethylaminoethylchloride hydrochloride

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H300-Fatal if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.

SDS Documents
Certificates