missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(Di-p-tolylphosphino)benzaldehyde, 97%

Brand:  Alfa Aesar™ H63183.03

Code : NEW

Additional Details : CAS Number : 1202865-03-9 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15409738

  • £60.90 / 1g


Estimated Shipment date
from Supplier 11-05-2018
Add to basket

Specifications

Chemical Identifiers

2-(Di-p-tolylphosphino)benzaldehyde
318.12
2-[Bis(4-methylphenyl)phosphino]benzaldehyde
1202865-03-9
MFCD15143661

Specifications

2-(Di-p-tolylphosphino)benzaldehyde
Yellow
1g
Solid
318.12
2-[Bis(4-methylphenyl)phosphino]benzaldehyde
Air Sensitive
105° to 111°C
1202865-03-9
97%
MFCD15143661
Videos
SDS