missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2'-Deoxycytidine, 99+%

Catalog Number 10076373
Click to view available options
Quantity:
250 mg
500 mg
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
951-77-9
C9H13N3O4
227.22
MFCD00006547
CKTSBUTUHBMZGZ-JORGKRSHNA-N
2'-deoxycytidine, deoxycytidine, dcyd, cytosine deoxyriboside, cytidine, 2'-deoxy, deoxyribose cytidine, desoxycytidin german, 4-amino-1-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl pyrimidin-2 1h-one, cytosine, deoxyribonucleoside, d-cytidine
13711
CHEBI:15698
4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one
NC1=NC(=O)N(C=C1)[C@H]1C[C@H](O)[C@@H](CO)O1
This item is not returnable. View return policy
951-77-9
C9H13N3O4
227.22
MFCD00006547
CKTSBUTUHBMZGZ-JORGKRSHNA-N
2'-deoxycytidine, deoxycytidine, dcyd, cytosine deoxyriboside, cytidine, 2'-deoxy, deoxyribose cytidine, desoxycytidin german, 4-amino-1-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl pyrimidin-2 1h-one, cytosine, deoxyribonucleoside, d-cytidine
13711
CHEBI:15698
4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one
NC1=NC(=O)N(C=C1)[C@H]1C[C@H](O)[C@@H](CO)O1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 99.0
CAS Max % 100.0
Melting Point 209°C to 211°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility in water: soluble
Formula Weight 227.22
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material 2'-Deoxycytidine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'