GE Healthcare 2'-Deoxyadenosine 5'-Triphosphate Solution


Overview
Brand: GE Healthcare

Manufacturer Part Number: 28406513

UNSPSC: 41106305

Code: GH

Additional Details:
Additional Details: Weight: 0.01400kg



Product Code. 10545145

Quantity Price
1 £ 378.46000000000004 / Each
Estimated Shipment
11-05-2017
Add to basket

If this item is out of stock, delivery will be in 3-4 working days providing the item is in stock at the supplier.
Please enquire with customer services on 01509 555500 for stock availability.

 

Description and Specification

Specification

Product Type 2'-Deoxycytidine 5'-Triphosphate Solution
Concentration 100mm
Purity > 99% Triphosphate
Format Liquid
Color Colorless
For Use With (Application) For Long PCR and Sequencing

By incorporating recent advances in manufacturing technology, GE Healthcare now offers illustra Solution dNTPs: the highest purity deoxynucleotides available for amplification, dideoxy sequencing, labeling, mutagenesis, cDNA synthesis, and expression profiling. A Certificate of Analysis for each lot is available on request.
  • Free from DNase, RNase, and nicking enzyme activity
  • Greater than 99% triphosphate purity (by HPLC) for optimum performance and consistency
  • Buffer free and ready-to-use solutions in multiple formats
  • Functionally tested for long PCR and sequencing
  • Custom formulation, testing, packaging, and labeling are available