missing translation for 'onlineSavingsMsg'
Learn More

2-Decanol, 98%, ACROS Organics™

Brand:  Acros Organics 331600250

Code : 29

Additional Details : CAS Number : 1120-06-5 Weight : 0.07500kg

Quantity 25g

 View more versions of this product

Product Code. 10623142

  • £44.40 / 25g


EU Stock 1
Add to basket

Specifications

Chemical Identifiers

2-Decanol
1120-06-5
100.0
98%
CH3(CH2)7CH(OH)CH3
MFCD00004594
95%
97.5
≥97.5%
C10H22O
158.28

Specifications

2-Decanol
1120-06-5
100.0
Glass bottle
Liquid
98%
CH3(CH2)7CH(OH)CH3
MFCD00004594
01, I, 213
95%
97.5
Authentic
1.4330 to 1.4350 (20°C, 589nm)
≥97.5%
C10H22O
158.28
Clear colorless
25g
Videos
SDS