missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Cyclopropyl-L-glycine, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
2-Cyclopropyl-L-glycine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Cyclopropyl-L-glycine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in water. Insoluble in ethanol.
Notes
Store at 4°C. Incompatible with heat and oxidizing agents. Protect from heat.
2-Cyclopropyl-L-glycine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in water. Insoluble in ethanol.
Notes
Store at 4°C. Incompatible with heat and oxidizing agents. Protect from heat.
Specifications
Color | White |
Beilstein | 5728866 |
Solubility Information | Soluble in water. Insoluble in ethanol. |
IUPAC Name | (2S)-2-azaniumyl-2-cyclopropylacetate |
Formula Weight | 115.13 |
Percent Purity | 97% |
Quantity | 250 mg |
Physical Form | Crystalline |
Chemical Name or Material | 2-Cyclopropyl-L-glycine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'