missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Cyclopropyl-L-glycine, 97%

Brand:  Alfa Aesar™ H65755.MD

Code : NEW

Additional Details : CAS Number : 49606-99-7 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15412629

  • £33.10 / 250mg


Estimated Shipment date
from Supplier 08-06-2018
Add to basket

Description

Description

Applications
2-Cyclopropyl-L-glycine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in water. Insoluble in ethanol.

Notes
Store at 4°C. Incompatible with heat and oxidizing agents. Protect from heat.
Specifications

Chemical Identifiers

2-Cyclopropyl-L-glycine
115.13
5728866
49606-99-7
MFCD06659116
(S)-2-Amino-2-cyclopropylacetic acid

Specifications

2-Cyclopropyl-L-glycine
Crystalline
115.13
5728866
White to off-white
49606-99-7
97%
MFCD06659116
(S)-2-Amino-2-cyclopropylacetic acid
250mg
Videos
SDS