missing translation for 'onlineSavingsMsg'
Learn More

2-Cyclohexen-1-ol, 95%, Thermo Scientific Chemicals

Catalog Number p-4228562
Click to view available options
Quantity:
1 g
5 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
822-67-3
C6H10O
98.14
MFCD00001570
PQANGXXSEABURG-UHFFFAOYSA-N
2-cyclohexen-1-ol, cyclohex-2-enol, 2-cyclohexene-1-ol, 2-cyclohexenol, 3-hydroxycyclohexene, 1-cyclohexen-3-ol, cyclohexen-3-ol, cyclohex-2-ene-1-ol, 1,2,3,4-tetrahydrophenol, cyclohexen-3-ol van
13198
cyclohex-2-en-1-ol
C1CC=CC(C1)O
This item is not returnable. View return policy
822-67-3
C6H10O
98.14
MFCD00001570
PQANGXXSEABURG-UHFFFAOYSA-N
2-cyclohexen-1-ol, cyclohex-2-enol, 2-cyclohexene-1-ol, 2-cyclohexenol, 3-hydroxycyclohexene, 1-cyclohexen-3-ol, cyclohexen-3-ol, cyclohex-2-ene-1-ol, 1,2,3,4-tetrahydrophenol, cyclohexen-3-ol van
13198
cyclohex-2-en-1-ol
C1CC=CC(C1)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Color Colorless
Density 0.9840g/mL
Boiling Point 164.0°C to 166.0°C
Flash Point 58°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula C6H9OH
Refractive Index 1.4855
Quantity 1 g
Beilstein 06,II,6
Specific Gravity 0.984
Formula Weight 98.14
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 2-Cyclohexen-1-ol
Show More Show Less
compliance-icons
Product Identifier
  • 2-Cyclohexen-1-ol
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'