missing translation for 'onlineSavingsMsg'
Learn More

2-Cyclohepten-1-one, 90%

Product Code. 15463534 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15463534

Brand: Thermo Scientific Acros 171070010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1121-66-0 | C7H10O | 110.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1121-66-0
Molecular Formula C7H10O
Molecular Weight (g/mol) 110.16
InChI Key WZCRDVTWUYLPTR-UHFFFAOYSA-N
Synonym 2-cyclohepten-1-one, cyclohept-2-enone, tropilene, 2-cycloheptenone, cycloheptenone, z-cyclohept-2-enone, cyclohept-2-eneone, cyclohept-en-1-one, acmc-1bphl, 2-cyclohepten-1-one, z
PubChem CID 70723
IUPAC Name cyclohept-2-en-1-one
SMILES C1CCC(=O)C=CC1

Specifications

Density 0.9900g/mL
Boiling Point 189.0°C
Flash Point 68°C
Infrared Spectrum Authentic
Assay Percent Range 89% min. (GC)
Packaging Glass Bottle
Linear Formula C7H10(=O)
Refractive Index 1.495
Quantity 1 g
Specific Gravity 0.99
Formula Weight 110.16
Percent Purity 90%
Chemical Name or Material 2-Cyclohepten-1-one
Show More Show Less
compliance-icons
Product Identifier
  • 2-Cyclohepten-1-one, 90%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.