missing translation for 'onlineSavingsMsg'
Learn More

2-Cyanothioacetamide, 98%, Thermo Scientific Chemicals

Catalog Number p-7022588
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
7357-70-2
C3H4N2S
100.14
MFCD00010025
BHPYMZQTCPRLNR-UHFFFAOYSA-N
2-cyanothioacetamide, cyanothioacetamide, ethanethioamide, 2-cyano, cyano thioacetamide, 2-cyano-thioacetamide, 3-amino-3-thioxopropanenitrile, cyanthioacetamide, cyanoacetothioamide, 2-cyanothioactamide, alpha-cyanothioacetamide
1416277
2-cyanoethanethioamide
NC(=S)CC#N
This item is not returnable. View return policy
7357-70-2
C3H4N2S
100.14
MFCD00010025
BHPYMZQTCPRLNR-UHFFFAOYSA-N
2-cyanothioacetamide, cyanothioacetamide, ethanethioamide, 2-cyano, cyano thioacetamide, 2-cyano-thioacetamide, 3-amino-3-thioxopropanenitrile, cyanthioacetamide, cyanoacetothioamide, 2-cyanothioactamide, alpha-cyanothioacetamide
1416277
2-cyanoethanethioamide
NC(=S)CC#N

2-Cyanothioacetamide was used in the synthesis of 3,4-trans-4-aryl-3-(1-pyridinio)-1,2,3,4-tetrahydropyridine-6-thiolates and bis[6-(2-aryl)-2-thioxo-1,2-dihydropyridine-3-carbonitrile]. 2-Cyanothioacetamide is used as a building block for 2-pyridothiones. It can be used in agrochemical, pharmaceutical and dyestuff field etc,.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Cyanothioacetamide was used in the synthesis of 3,4-trans-4-aryl-3-(1-pyridinio)-1,2,3,4-tetrahydropyridine-6-thiolates and bis[6-(2-aryl)-2-thioxo-1,2-dihydropyridine-3-carbonitrile]. 2-Cyanothioacetamide is used as a building block for 2-pyridothiones. It can be used in agrochemical, pharmaceutical and dyestuff field etc,.

Solubility
Does not mix well with water.

Notes
Light Sensitive. Store in the dark.
TRUSTED_SUSTAINABILITY
Melting Point 114°C to 120°C
Quantity 5 g
UN Number UN3439
Beilstein 1699934
Sensitivity Light sensitive
Solubility Information Does not mix well with water.
Formula Weight 100.14
Percent Purity 98%
Chemical Name or Material 2-Cyanothioacetamide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'