missing translation for 'onlineSavingsMsg'
Learn More

2-Cyanomethyl-4-(4-hydroxyphenyl)thiazole, 97%, Thermo Scientific™

Catalog Number p-7039082
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
1028352-86-4
C11H8N2OS
216.258
MFCD22682948
CMIGLFUTBISGQV-UHFFFAOYSA-N
2-cyanomethyl-4-4-hydroxyphenyl thiazole, 2-4-4-hydroxyphenyl-1,3-thiazol-2-yl acetonitrile
73996315
2-[4-(4-hydroxyphenyl)-1,3-thiazol-2-yl]acetonitrile
C1=CC(=CC=C1C2=CSC(=N2)CC#N)O
This item is not returnable. View return policy
1028352-86-4
C11H8N2OS
216.258
MFCD22682948
CMIGLFUTBISGQV-UHFFFAOYSA-N
2-cyanomethyl-4-4-hydroxyphenyl thiazole, 2-4-4-hydroxyphenyl-1,3-thiazol-2-yl acetonitrile
73996315
2-[4-(4-hydroxyphenyl)-1,3-thiazol-2-yl]acetonitrile
C1=CC(=CC=C1C2=CSC(=N2)CC#N)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 145°C to 151°C
Quantity 2 g
UN Number UN3439
Formula Weight 216.25
Percent Purity 97%
Chemical Name or Material 2-Cyanomethyl-4-(4-hydroxyphenyl)thiazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'