missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Cyanoethyl-N,N,N',N'-tetraisopropylphosphorodiamidite, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Density | 0.9490g/mL |
Boiling Point | 98.0°C to 100.0°C (5.0 mmHg) |
Flash Point | 60°C |
Assay Percent Range | 95% |
Packaging | Glass bottle |
Refractive Index | 1.47 |
Quantity | 25 g |
Specific Gravity | 0.949 |
Formula Weight | 301.41 |
Percent Purity | 95% |
Show More |
Safety and Handling
Product Identifier
- 2-Cyanoethyl-N,N,N',N'-tetraisopropylphosphorodiamidite
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Flammable liquid Category 3
- Skin sensitiser Category 1
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H317-May cause an allergic skin reaction.
- H412-Harmful to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'