Learn More
2-cyanoethyl N,n-diisopropylchlorophosphoramidite, 97%, ACROS Organics™
Brand: Acros Organics 374830050
Code : 29
Additional Details : CAS Number : 89992-70-1 Weight : 0.05500kg Transport : UN number : 2845 Chem class : 1 Pack group : I
Quantity | 5g |
---|---|
Packaging | Glass bottle |
Chemical Identifiers
2-Cyanoethyl N,N-diisopropylchlorophosphoramidite | |
96.0 | |
97% | |
C9H18ClN2OP | |
236.68 |
89992-70-1 | |
100.0 | |
97% | |
[(CH3)2CH]2NP(Cl)O(CH2)2CN | |
MFCD00011544 |
Specifications
2-Cyanoethyl N,N-diisopropylchlorophosphoramidite | |
5g | |
89992-70-1 | |
100.0 | |
96.0% min. | |
Glass bottle | |
97% | |
1.4760 to 1.4800 | |
[(CH3)2CH]2NP(Cl)O(CH2)2CN | |
MFCD00011544 | |
White to yellow |
110°C | |
May develop turbidity on storage | |
96.0 | |
1.0610g/mL | |
Authentic | |
Crystalline Powder | |
97% | |
C9H18ClN2OP | |
236.68 | |
Solubility in water: reacts. |