missing translation for 'onlineSavingsMsg'
Learn More

2-Cyanobenzoic acid, 94%, Thermo Scientific Chemicals

Catalog Number p-3770853
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
3839-22-3
C8H5NO2
147.13
MFCD00045796
DTNSDCJFTHMDAK-UHFFFAOYSA-N
o-cyanobenzoic acid, cyanobenzoic acid, benzoic acid, 2-cyano, 2-carboxybenzonitrile, 2-carboxy-benzonitrile, 2-cyanobenzoicacid, 2-cyano-benzoic acid, pubchem12513, acmc-1bn2v
138061
2-cyanobenzoic acid
C1=CC=C(C(=C1)C#N)C(=O)O
This item is not returnable. View return policy
3839-22-3
C8H5NO2
147.13
MFCD00045796
DTNSDCJFTHMDAK-UHFFFAOYSA-N
o-cyanobenzoic acid, cyanobenzoic acid, benzoic acid, 2-cyano, 2-carboxybenzonitrile, 2-carboxy-benzonitrile, 2-cyanobenzoicacid, 2-cyano-benzoic acid, pubchem12513, acmc-1bn2v
138061
2-cyanobenzoic acid
C1=CC=C(C(=C1)C#N)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 92.0
CAS Max % 100.0
Melting Point 212.0°C
Color Colorless to Yellow or White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 92% min. (HPLC)
Packaging Glass bottle
Linear Formula NCC6H4CO2H
Quantity 1 g
Formula Weight 147.13
Percent Purity 94%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 2-Cyanobenzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Cyanobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'