missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Cyano-5-(trifluoromethyl)pyridine, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
It is a useful reactant in the synthesis of s-tetrazines. Also act as a intermediate for pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is a useful reactant in the synthesis of s-tetrazines. Also act as a intermediate for pharmaceutical.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
It is a useful reactant in the synthesis of s-tetrazines. Also act as a intermediate for pharmaceutical.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
Quantity | 250 mg |
UN Number | UN3439 |
Beilstein | 13339826 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 172.11 |
Percent Purity | 98% |
Physical Form | Solid |
Chemical Name or Material | 2-Cyano-5-(trifluoromethyl)pyridine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'