missing translation for 'onlineSavingsMsg'
Learn More

2-Cyano-3-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Catalog Number 15477336
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
765916-91-4
C13H15BFNO2
247.076
TXGKJULMHJEZTA-UHFFFAOYSA-N
2-fluoro-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 2-cyano-3-fluorophenylboronic acid pinacol ester, benzonitrile, 2-fluoro-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 2-cyano-3-fluorophenylboronic acid, pinacol ester, 2-cyano-3-fluorobenzeneboronic acid pinacol ester, 2-fluoro-6-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 6-fluoro-2-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzonitrile
22273603
2-fluoro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
B1(OC(C(O1)(C)C)(C)C)C2=C(C(=CC=C2)F)C#N
This item is not returnable. View return policy
765916-91-4
C13H15BFNO2
247.076
TXGKJULMHJEZTA-UHFFFAOYSA-N
2-fluoro-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 2-cyano-3-fluorophenylboronic acid pinacol ester, benzonitrile, 2-fluoro-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 2-cyano-3-fluorophenylboronic acid, pinacol ester, 2-cyano-3-fluorobenzeneboronic acid pinacol ester, 2-fluoro-6-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 6-fluoro-2-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzonitrile
22273603
2-fluoro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
B1(OC(C(O1)(C)C)(C)C)C2=C(C(=CC=C2)F)C#N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 247.07
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 2-Cyano-3-fluorobenzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'