missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Cyano-2-(methoxyimino)acetamidine, 97%

Brand:  Alfa Aesar™ H61847.14

Code : NEW

Additional Details : CAS Number : 84981-64-6 Weight : 0.02500kg Transport : UN number : 3439 Chem class : 1 Pack group : III

Quantity 25g

 View more versions of this product

Product Code. 15418438

  • £217.80 / 25g


Estimated Shipment date
from Supplier 19-02-2018
Add to basket

Specifications

Chemical Identifiers

2-Cyano-2-(methoxyimino)acetamidine
84981-64-6
126.12
UN3439

Specifications

2-Cyano-2-(methoxyimino)acetamidine
84981-64-6
97%
UN3439
25g
Solid
126.12
Videos
SDS