missing translation for 'onlineSavingsMsg'
Learn More

2-Cl-IB-MECA, Tocris Bioscience™

Highly selective A3 agonist

Brand:  Tocris (Bio-Techne) 1104/10

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13488493

  • £302.29 / 10mg


Estimated Shipment
03-10-2018
Add to basket

Description

Description

High affinity and extremely selective A3 adenosine receptor agonist (Ki = 0.33 nM). Displays 2500- and 1400-fold selectivity over A1 and A2A receptors respectively. Exhibits high selectivity over the Na+-independent adenosine transporter.
Specifications

Chemical Identifiers

163042-96-4
544.734
1-[2-Chloro-6-[[(3-iodophenyl)methyl]amino]-9H-purin-9-yl]-1-deoxy-N-methyl-β-D-ribofuranuronamide
C18H18ClIN6O4
IPSYPUKKXMNCNQ-PFHKOEEOSA-N
3035850

Specifications

2-Cl-IB-MECA
163042-96-4
C18H18ClIN6O4
IPSYPUKKXMNCNQ-PFHKOEEOSA-N
(2S,3S,4R,5R)-5-[2-chloro-6-[(3-iodophenyl)methylamino]purin-9-yl]-3,4-dihydroxy-N-methyloxolane-2-carboxamide
3035850
10mg
>98%
1-[2-Chloro-6-[[(3-iodophenyl)methyl]amino]-9H-purin-9-yl]-1-deoxy-N-methyl-β-D-ribofuranuronamide
CNC(=O)C1C(C(C(O1)N2C=NC3=C2N=C(N=C3NCC4=CC(=CC=C4)I)Cl)O)O
544.734
544.74
Videos