missing translation for 'onlineSavingsMsg'
Learn More

2-chlorothiophenol, 98%, Thermo Scientific™

Catalog Number p-3757289
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
6320-03-2
C6H5ClS
144.62
MFCD00004830
PWOBDMNCYMQTCE-UHFFFAOYSA-N
This item is not returnable. View return policy
6320-03-2
C6H5ClS
144.62
MFCD00004830
PWOBDMNCYMQTCE-UHFFFAOYSA-N
CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Density 1.2750g/mL
Flash Point 88°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula ClC6H4SH
Refractive Index 1.6020 to 1.6040 (20°C, 589nm)
Quantity 5 g
Specific Gravity 1.275
Synonym 2-chlorothiophenol, o-chlorothiophenol, benzenethiol, 2-chloro, 1-chloro-2-mercaptobenzene, o-chlorobenzenethiol, o-chloromercaptobenzene, chlorothiophenol, benzenethiol, o-chloro, 2-chlorobenzene-1-thiol, 2-chloro thiophenol
SMILES C1=CC=C(C(=C1)S)Cl
IUPAC Name 2-chlorobenzenethiol
PubChem CID 80599
Formula Weight 144.62
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-chlorothiophenol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Chlorothiophenol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'