missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorothiophene, 98+%, Thermo Scientific™

Catalog Number p-7080735
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy
96-43-5
C4H3ClS
118.578
MFCD00005421
GSFNQBFZFXUTBN-UHFFFAOYSA-N
2-thienyl chloride, thiophene, 2-chloro, 2-chloro thiophene, 2-chlorthiophen, chlorothiophene, thienyl chloride, 5-chlorothiophene, thiophene, chloro, sfpdadhhryszzp@, pubchem7376
7299
2-chlorothiophene
C1=CSC(=C1)Cl
This item is not returnable. View return policy
96-43-5
C4H3ClS
118.578
MFCD00005421
GSFNQBFZFXUTBN-UHFFFAOYSA-N
2-thienyl chloride, thiophene, 2-chloro, 2-chloro thiophene, 2-chlorthiophen, chlorothiophene, thienyl chloride, 5-chlorothiophene, thiophene, chloro, sfpdadhhryszzp@, pubchem7376
7299
2-chlorothiophene
C1=CSC(=C1)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used in the preparation of 5-phenylthiophene derivative and N-arylhydroxamic acids.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from electrostatic charges and keep away from ignition sources.
TRUSTED_SUSTAINABILITY
Melting Point -72°C
Density 1.285
Boiling Point 127°C to 129°C
Flash Point 22°C (71°F)
Odor Unpleasant
Refractive Index 1.548
Quantity 50 g
UN Number UN1992
Beilstein 104652
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 118.59
Percent Purity ≥98%
Chemical Name or Material 2-Chlorothiophene
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'