missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chlorothiophene, 97%, Thermo Scientific™
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Specifications
CAS Min % | 96.0 |
CAS Max % | 100.0 |
Melting Point | -71.9°C |
Color | Colorless to Brown |
Density | 1.2800g/mL |
Boiling Point | 127.0°C to 129.0°C |
Flash Point | 22°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 96% min. (GC) |
Packaging | Glass bottle |
Show More |
Safety and Handling
Product Identifier
- 2-Chlorothiophene
- Danger
- Acute toxicity Category 3
- Flammable liquid Category 2
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'