missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorothiophene, 97%, Thermo Scientific™

Catalog Number 10489260
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
96-43-5
C4H3ClS
118.58
MFCD00005421
GSFNQBFZFXUTBN-UHFFFAOYSA-N
2-thienyl chloride, thiophene, 2-chloro, 2-chloro thiophene, 2-chlorthiophen, chlorothiophene, thienyl chloride, 5-chlorothiophene, thiophene, chloro, sfpdadhhryszzp@, pubchem7376
7299
2-chlorothiophene
C1=CSC(=C1)Cl
This item is not returnable. View return policy
96-43-5
C4H3ClS
118.58
MFCD00005421
GSFNQBFZFXUTBN-UHFFFAOYSA-N
2-thienyl chloride, thiophene, 2-chloro, 2-chloro thiophene, 2-chlorthiophen, chlorothiophene, thienyl chloride, 5-chlorothiophene, thiophene, chloro, sfpdadhhryszzp@, pubchem7376
7299
2-chlorothiophene
C1=CSC(=C1)Cl
CAS Min % 96.0
CAS Max % 100.0
Melting Point -71.9°C
Color Colorless to Brown
Density 1.2800g/mL
Boiling Point 127.0°C to 129.0°C
Flash Point 22°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5470 to 1.5490
Quantity 100 mL
Beilstein 17, 32
Specific Gravity 1.28
Solubility Information Solubility in water: insoluble in water. Other solubilities: miscible with ethanol, ether, slightly soluble in chloroform
Formula Weight 118.58
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Chlorothiophene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Chlorothiophene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'