missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorothiophene, 97%, ACROS Organics™

Brand:  Acros Organics 168265000

Code : 29

Additional Details : CAS Number : 96-43-5 Weight : 0.69000kg Transport : UN number : 1992 Chem class : 1 Pack group : II


Quantity 500mL
Packaging Glass bottle

 View more versions of this product

Product Code. 10197403

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

2-Chlorothiophene
96.0
≥96.0%
C4H3ClS
MFCD00005421
96-43-5
100.0
97%
118.58

Specifications

2-Chlorothiophene
Clear colorless to light brown
96-43-5
100.0
Glass bottle
1.5470 to 1.5490 (20°C, 589nm)
≥96.0%
C4H3ClS
MFCD00005421
1.28g/mL
500mL
96.0
Authentic
Product darkens on storage or exposure to light
Liquid
97%
118.58
17, 32
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Chlorothiophene

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2

Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H225-Highly flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor / A§
  • P302+P352-IF ON SKIN: Wash with plenty of water/A§.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/A§/if you feel unwell.

SDS