missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Chloropyridine-4-carboxamidoxime, 97%

Brand:  Alfa Aesar™ H65100.03

Code : NEW

Additional Details : CAS Number : 857653-85-1 Weight : 0.00100kg Transport : UN number : 2811 Chem class : 1 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 15422469

  • £84.16 / 1g


Estimated Shipment date
from Supplier 26-03-2018
Add to basket

Specifications

Chemical Identifiers

2-Chloropyridine-4-carboxamidoxime
171.59
UN2811
857653-85-1
MFCD11501096
2-Chloro-N-hydroxyisonicotinamidine

Specifications

2-Chloropyridine-4-carboxamidoxime
160° to 166°C
857653-85-1
97%
MFCD11501096
2-Chloro-N-hydroxyisonicotinamidine
White to pale brown
1g
Solid
171.59
UN2811
Videos
SDS