missing translation for 'onlineSavingsMsg'
Learn More

2-chloropyridine-3-sulfonyl chloride, 97%, ACROS Organics™

Brand:  Acros Organics 434310010

Code : 29

Additional Details : CAS Number : 6684-06-6 Weight : 0.05100kg Transport : UN number : 3261 Chem class : 1a Pack group : III


Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 11936361

  • £111.30 / 1g


Estimated Shipment
16-03-2018
Add to basket


Specifications

Chemical Identifiers

2-Chloropyridine-3-sulfonyl chloride
96.0
≥96.0% diethylsulfonamide derivative (HPLC)
C5H3Cl2NO2S
6684-06-6
100.0
97%
212.06

Specifications

2-Chloropyridine-3-sulfonyl chloride
42.0° to 53.0°C
6684-06-6
100.0
Glass bottle
≥96.0% diethylsulfonamide derivative (HPLC)
C5H3Cl2NO2S
White to off-white
1g
96.0
Authentic
Powder
97%
212.06
Videos
SDS