missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Chloropyridine-3-carboxamidoxime, 97%

Brand:  Alfa Aesar™ H65400.03

Code : NEW

Additional Details : CAS Number : 468068-58-8 Weight : 0.00100kg Transport : UN number : 2811 Chem class : 1 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 15412549

  • £73.67 / 1g


Estimated Shipment date
from Supplier 19-03-2018
Add to basket

Specifications

Chemical Identifiers

2-Chloropyridine-3-carboxamidoxime
171.59
UN2811
468068-58-8
MFCD08273500
2-Chloro-N-hydroxynicotinamidine

Specifications

2-Chloropyridine-3-carboxamidoxime
155° to 161°C
468068-58-8
97%
MFCD08273500
2-Chloro-N-hydroxynicotinamidine
White to cream
1g
Solid
171.59
UN2811
Videos
SDS