missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chlorophenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
2-Chlorophenylhydrazine hydrochloride is used to study intrinsic resistance of Mycobacterium smegmatis to fluoroquinolones, used for treatment of Mycobacterium tuberculosis. It may be used in pyrazoline synthesis. It can be used to produce N-azepan-2-ylidene-N'-(2-chloro-phenyl)-hydrazine with 7-methoxy-3,4,5,6-tetrahydro-2H-azepine at heating.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Chlorophenylhydrazine hydrochloride is used to study intrinsic resistance of Mycobacterium smegmatis to fluoroquinolones, used for treatment of Mycobacterium tuberculosis. It may be used in pyrazoline synthesis. It can be used to produce N-azepan-2-ylidene-N′-(2-chloro-phenyl)-hydrazine with 7-methoxy-3,4,5,6-tetrahydro-2H-azepine at heating.
Solubility
Soluble in water.
Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
2-Chlorophenylhydrazine hydrochloride is used to study intrinsic resistance of Mycobacterium smegmatis to fluoroquinolones, used for treatment of Mycobacterium tuberculosis. It may be used in pyrazoline synthesis. It can be used to produce N-azepan-2-ylidene-N′-(2-chloro-phenyl)-hydrazine with 7-methoxy-3,4,5,6-tetrahydro-2H-azepine at heating.
Solubility
Soluble in water.
Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
Specifications
Melting Point | ∼201°C (decomposition) |
Quantity | 5 g |
Beilstein | 3699381 |
Solubility Information | Soluble in water. |
Formula Weight | 179.05 |
Percent Purity | 97% |
Chemical Name or Material | 2-Chlorophenylhydrazine hydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'