missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorophenylboronic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-2698185
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
3900-89-8
C6H6BClO2
156.37
MFCD00674012
RRCMGJCFMJBHQC-UHFFFAOYSA-N
2-chlorophenyl boronic acid, 2-chlorobenzeneboronic acid, 2-chlorophenylboronicacid, 2-chlorophenylbornic acid, o-chlorophenylboronic acid, 2-chlorophenyl boranediol, boronic acid, 2-chlorophenyl, 2-chloro-phenyl-boronic acid, boronic acid, b-2-chlorophenyl
2734322
(2-chlorophenyl)boronic acid
OB(O)C1=CC=CC=C1Cl
This item is not returnable. View return policy
3900-89-8
C6H6BClO2
156.37
MFCD00674012
RRCMGJCFMJBHQC-UHFFFAOYSA-N
2-chlorophenyl boronic acid, 2-chlorobenzeneboronic acid, 2-chlorophenylboronicacid, 2-chlorophenylbornic acid, o-chlorophenylboronic acid, 2-chlorophenyl boranediol, boronic acid, 2-chlorophenyl, 2-chloro-phenyl-boronic acid, boronic acid, b-2-chlorophenyl
2734322
(2-chlorophenyl)boronic acid
OB(O)C1=CC=CC=C1Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 94.0°C to 95.0°C
CAS Min % 96.0
Color White
CAS Max % 100.0
Quantity 1 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Powder
Assay Percent Range 96% min. (HPLC)
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ether,thf,dmso,dmf,methanol
Formula Weight 156.38
Percent Purity 97%
Chemical Name or Material 2-Chlorophenylboronic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'