Learn More
2-Chlorophenylboronic acid, 97%, ACROS Organics™
Brand: Acros Organics 358990050
Code : 29
Additional Details : CAS Number : 3900-89-8 Weight : 0.05500kg
Quantity | 5g |
---|---|
Packaging | Glass bottle |
Chemical Identifiers
2-Chlorophenylboronic acid | |
96.0 | |
97% | |
C6H6BClO2 | |
MFCD00674012 |
3900-89-8 | |
100.0 | |
97% | |
156.38 |
Specifications
2-Chlorophenylboronic acid | |
94.0° to 95.0°C | |
3900-89-8 | |
100.0 | |
Glass bottle | |
97% | |
C6H6BClO2 | |
MFCD00674012 |
White | |
5g | |
96.0 | |
Authentic | |
Crystalline Powder | |
97% | |
156.38 |